L(+)-Serine cell culture reagent
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine
J62187.09 J62187.22 J62187.36 J62187.A1
J62187.09EA 35.4 EUR
J62187.09 J62187.22 J62187.36 J62187.A1
L(+)-Serine cell culture reagent
L(+)-Serine
CAS No.: 56-45-1 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Physical Form: Powder Appearance: White Melting Point: 222° Beilstein Reference: 1721404 Merck Reference: 14,8460 MDL No.: MFCD00064224
Formule: C₃H₇NO₃
Kookpunt: 259 °C (1013 hPa)
Smeltpunt: 222 °C
Dichtheid: 1,6 g/cm³ (22 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064224
CAS nummer: 56-45-1
EINECS: 200-274-3
Merck Index: 14,8460