L-Lysine ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J62225.14
J62225.22
J62225.36
J62225.14EA
85.4
EUR
J62225.14
J62225.22
J62225.36
L-Lysine ≥98%
L-Lysine
Purity: 98%
CAS No.: 56-87-1
Molecular Formula: C6H14N2O2
Formula Weight: 146.19
Physical Form: Powder
Appearance: White
Melting Point: 215°
Solubility: Soluble in water
Beilstein Reference: 1722531
Merck Reference: 14,5636
Optical Rotation: +26° (c=2 in 6M HCl)
MDL No.: MFCD00064433
Formule:
C₆H₁₄N₂O₂ Kookpunt: 290 °C (1013 hPa) Smeltpunt: 215 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00151035 CAS nummer: 56-87-1 EINECS: 200-294-2 Merck Index: 14,5636 |