Dextran, M.W. ~ 150,000
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J63789.09 J63789.18 J63789.22 J63789.36 J63789.A1
J63789.09EA 37.7 EUR
J63789.09 J63789.18 J63789.22 J63789.36 J63789.A1
Dextran, M.W. ~ 150,000
Dextran
CAS No.: 9004-54-0 Molecular Formula: (C6H16O5)n Physical Form: Powder Appearance: White Merck Reference: 14,2948 MDL No.: MFCD00130935
Formule: (C₆H₁₀O₅)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00130935
CAS nummer: 9004-54-0
EINECS: 232-677-5
Merck Index: 14,2948