Magnesium chloride 1 M in aqueous solution, sterile-filtered
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J61014.XE J61014.AK
J61014.XEEA 75 EUR
J61014.XE J61014.AK
Magnesium chloride 1 M in aqueous solution, sterile-filtered
Magnesium chloride
Formule: MgCl₂
MW: 95.21 g/mol
Kookpunt: 1412
Smeltpunt: 715
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011106
CAS nummer: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684