4-Fluorobenzaldehyde 98+%, AcroSeal®

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

427191000.EA 89.2 EUR
427191000.
4-Fluorobenzaldehyde 98+%, AcroSeal®
4-Fluorobenzaldehyde
Formule: C₇H₅FO
MW: 124,11 g/mol
Kookpunt: 181 °C (758 mmHg)
Smeltpunt: –10 °C
Dichtheid: 1,15 g/cm³
Vlampunt: 56 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003378
CAS nummer: 459-57-4
EINECS: 207-293-6
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5195 to 1.5215 (20°C, 589 nm)
GC ≥98.0 %
Residue after evaporation ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR