Chlortetracycline hydrochloride
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
CAS No.: 64-72-2
Molecular Formula: C22H23ClN2O8.HCl
Formula Weight: 515.34
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Yellow to brown
Melting Point: 210-215° dec.
Solubility: Soluble in water
Beilstein Reference: 3858364
Merck Reference: 14,2192
MDL No.: MFCD00082440
Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria
Formule:
C₂₂H₂₃ClN₂O₈·HCl MW: 515.34 g/mol Smeltpunt: 210…215 °C (ontbinding) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD26142699 CAS nummer: 64-72-2 EINECS: 200-591-7 |