Oxytetracycline hydrochloride

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

J62489.14 J62489.22
J62489.14EA 80 EUR
J62489.14 J62489.22
Oxytetracycline hydrochloride
Oxytetracycline hydrochloride

CAS No.: 2058-46-0 Molecular Formula: C22H24N2O9.HCl Formula Weight: 496.90 Physical Form: Crystalline powder Sensitivity: Air and Light Sensitive Appearance: Yellow to pale gray Melting Point: 180° Solubility: Soluble in ethanol Beilstein Reference: 3853107 Merck Reference: 14,6976 MDL No.: MFCD00135815


Impairs mitochondrial protein synthesis

Formule: C₂₂H₂₅ClN₂O₉
Smeltpunt: 180 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00135815
CAS nummer: 2058-46-0
EINECS: 218-161-2
Merck Index: 14,6976

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR