N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J60173.03
J60173.06
J60173.03EA
107
EUR
J60173.03
J60173.06
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
CAS No.: 85261-20-7
Molecular Formula: C17H35NO6
Formula Weight: 349.46
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 91-93°
Solubility: Soluble in methanol at 50mg/ml. Also soluble in water at 10mg/ml 20 °C
Beilstein Reference: 6974359
MDL No.: MFCD00036801
Non-ionic detergent for solubilizing membrane proteins
Formule:
C₁₇H₃₅NO₆ Smeltpunt: 91…93 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00036801 CAS nummer: 85261-20-7 |