Auramine O ≥80%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow
J60343.14 J60343.18
J60343.14EA 37.6 EUR
J60343.14 J60343.18
Auramine O ≥80%
Auramine O
Formule: C₁₇H₂₂ClN₃
MW: 303.84 g/mol
Smeltpunt: 250 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012484
CAS nummer: 2465-27-2
EINECS: 219-567-2
UN: 2811
ADR: 6.1 III