Alcian blue 8GX
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
J60122.09 J60122.14 J60122.22
J60122.09EA 69.2 EUR
J60122.09 J60122.14 J60122.22
Alcian blue 8GX
Alcian blue
CAS No.: 33864-99-2 Molecular Formula: C56H68Cl4CuN16S4 Formula Weight: 1298.88 Physical Form: Powder Appearance: Dark blue Melting Point: 148° MDL No.: MFCD00010720
Formule: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
Smeltpunt: 148 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010720
CAS nummer: 33864-99-2
EINECS: 251-705-7
Merck Index: 14,218