Diphenyleneiodonium chloride

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

J64838.MB J64838.MF J64838.MC
J64838.MBEA 0 EUR
J64838.MB J64838.MF J64838.MC
Diphenyleneiodonium chloride
Diphenyleneiodonium chloride
Formule: C₁₂H₈ClI
Smeltpunt: >310 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00214165
CAS nummer: 4673-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR