Pipette stand, 337
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
320.337B 320.337G 320.337M 320.337Y 320.337R
613-4830EA 135 EUR
613-4830 613-4831 613-0546 613-4832 SOCO320.337R
Pipette stand, 337
Pipethouders
This space-saving pipette stand made from PA holds up to seven single channel pipettes of most brands.
  • Made of sturdy polyamid material, easy to clean
  • Very stable with anti-slip pads
  • Available in five pastel colours