Safety shoes, lace-up, Audrey
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JJD09/35 JJD09/36 JJD09/37 JJD09/38 JJD09/39 JJD09/40 JJD09/41
114-4336EA 0 EUR
114-4336 114-4337 114-4338 114-4339 114-4340 114-4341 114-4342
Safety shoes, lace-up, Audrey
Schoenen en laarzen
Laced shoes made from full grain leather, nylon lining and dual-density sole.
  • Antibacterial nylon lining
  • Full piece, anti-static and anatomic integrated insole, perforated in the front part for better foot venting, cushioning effect in the heel area
  • TPU/TPU versatile dual-density sole is resistant to slipping and abrasion
  • Protective steel toecap (200 J)
  • Light, metal-free composite puncture resistant midsole
Colour: Black
Certificaten: EN ISO 20345:2007; S3, SRC