Safety stand for BTX generator and BTX cuvettes plus
Leverancier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45-0207 45-0213
732-0034EA 1790 EUR
732-0034 BTXI45-0213
Safety stand for BTX generator and BTX cuvettes plus
Staafstatieven
The BTX safety stand is specially designed to connect to any BTX generator allowing for the safe delivery of HV electrical pulses to cuvettes. Up to two cuvettes may be electroporated in the safety stand simultaneously.