Erlenmeyer flasks, with Quickfit® ground joint
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1132/14 1132/06 1132/02 1132/20 1132/26 1132/15
BIBB1132/14EA 219 EUR
BIBB1132/14 BIBB1132/06 BIBB1132/02 BIBB1132/20 BIBB1132/26 BIBB1132/15
Erlenmeyer flasks, with Quickfit® ground joint
Kolven Erlenmeyers
Erlenmeyer flasks, made of Pyrex® borosilicate glass, with standard ground joint.
  • Resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Erlenmeyer flasks with high quality Quickfit® ground glass socket