Racks for chest freezers, Alu FlexRack
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TE20110 TE20112 TE20114 TE20116 TE20118 TE20120 TE20122 TE20124
TENATE20110EA 110.53 EUR
TENATE20110 TENATE20112 TENATE20114 TENATE20116 TENATE20118 TENATE20120 TENATE20122 TENATE20124
Racks for chest freezers, Alu FlexRack
Rekken Rekken voor cryogene dozen
Aluminium.
  • Adjustable shelves - the rack can be used for all or a mixture of cryobox heights
  • Can be used for any chest freezer
  • Only buy the additional shelves you need
The Alu FlexRack is recommended for all applications where a flexible storage solution is needed.
Leveringsinformatie: Supplied with locking rod for securing boxes and one shelf.