Automatic burettes, PTFE key, class AS, Schellbach
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3396/02AS 3396/04AS 3396/06AS
BIBB3396/02ASEA 289 EUR
BIBB3396/02AS BIBB3396/04AS BIBB3396/06AS
Automatic burettes, PTFE key, class AS, Schellbach
Buretten
Pyrex® borosilicate glass, class AS, with PTFE key stopcock of 1,5 mm bore.
  • Automatic zeroing
  • Schellbach stripe, with blue enamel graduations
Tube with vertical white stripe and central blue ribbon for accurate reading of the meniscus (where the blue ribbon splits into a wide and narrow band) and features a 29/32 Quickfit® cone.
Certificaten: Complies with ISO 385 and DIN 12700
Bestelinformatie: Burettes and reservoir bottles should be ordered separately.