Automatic burettes, with intermediate stopcock, class AS, Schellbach
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3496/02AS 3496/04AS 3496/06AS
BIBB3496/02ASEA 317 EUR
BIBB3496/02AS BIBB3496/04AS BIBB3496/06AS
Automatic burettes, with intermediate stopcock, class AS, Schellbach
Buretten
Burettes made of chemically resistant Pyrex® borosilicate glass and feature a 29/32 Quickfit® cone.
  • With automatic zero facility
  • Blue enamel graduations with Schellbach stripe
  • PTFE stopcock
Schellbach burette tube with vertical white stripe and central blue ribbon, facilitates accurate reading of the meniscus, also exact position of the meniscus is clearly defined where the blue ribbon splits into a wide and narrow band.