Pouring rings for laboratory bottles
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4536/32D 4536/45D
BIBB4536/32DEA 28.3 EUR
BIBB4536/32D BIBB4536/45D
Pouring rings for laboratory bottles
Sluitingen Flesdoppen
PYREX® ETFE.
  • For GL threads
  • Ideal for laboratory media bottles of high temperatures
  • Heat resistant up to +180 °C