Screw caps for laboratory/media bottles
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4526/25D 4526/32D 4526/45D
BIBB4526/25DEA 75.5 EUR
BIBB4526/25D BIBB4526/32D BIBB4526/45D
Screw caps for laboratory/media bottles
Sluitingen Flesdoppen
High temp red screw cap, made of PBT, for media-lab bottle.
  • Ideal for applications involving high temperatures or aggressive chemicals
  • With natural coloured PTFE faced sealing disc
  • Withstands hot air sterilisation up to 180 °C
  • PTFE sealing disc resists chemical attack