Spill pallet, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PAK604-BK-WOD PAK672-BK-WOD
NEWPPAK604-BK-WODEA 428 EUR
NEWPPAK604-BK-WOD NEWPPAK672-BK-WOD
Spill pallet, PIG®
Opslag voor vaten
These spill pallets are ideal for long-term storage of liquids with high specific gravities and molded-in sump catches leaks, drips and spills.
  • Sump made of linear LDPE with inhibitors and grate made of injection-molded PE foam
  • Innovative design transfers weight to load-bearing channels to maximise capacity
  • LDPE construction resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
  • Two-way forklift entry allows easy movement from either direction (loaded or unloaded)
Steel ramp fits two orMore Product Information