Proteome kits, mammalian proteome, FOCUS™
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-246EA 422 EUR
786-246
Proteome kits, mammalian proteome, FOCUS™
Eiwitextractie Protein Extraction Kits
Extracts and solubilises nearly all of the proteins from mammalian samples, including membrane as well as soluble proteins, by a strong chaotropic extraction buffer to solubilise even the most difficult proteins. Suitable for biological samples from animal tissues and adherent and suspension cells.
  • Single step extraction protocol
  • Supplied with a strong proprietary chaotropic extraction buffer
  • For 50×100 mg animal tissue or 50 μl wet cell pellet preparations
  • Suitable for sample preparation for 2D gel electrophoresis and other applications