Protein fractionation kit, FOCUS™ Membrane Proteins
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-249EA 958 EUR
786-249
Protein fractionation kit, FOCUS™ Membrane Proteins
Eiwitextractie Protein Extraction Kits
FOCUS™ Membrane Proteins for the fractionation of highly enriched membrane proteins.
  • Phase partition based extraction of membrane proteins
  • Fractions suitable for 2D electrophoresis and other applications
FOCUS™ Membrane Proteins is a rapid and highly reproducible method for preparation of membrane or hydrophobic proteins from biological samples for 2D-gel analysis or other applications. Membrane proteins are extracted with a single step phase partition, with an efficiency greater than 90% with minimal cross-contamination from hydrophilic proteins.
The kit is provided with reagents necessary for extraction of membrane proteins and their subsequent preparation for isoelectric focusing or 2D gel resolution for improved spot resolution. FOCUS™ Membrane Proteins kit is designed for >50 preps, where 1 prep is either 100 mg mammalian tissue, 50 µl wet animal cell pellet, 50 µl wet yeast pellet, 50 µl wet bacteria pellet, or 250 mg plant tissue.