Albumin removal kit, AlbuminOUT™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-251 786-252 786-251T
786-251EA 726 EUR
786-251 786-252 786-251T
Albumin removal kit, AlbuminOUT™
Eiwitextractie Protein Extraction Kits
Samples that contain a large abundance of albumin, such as plasma and cerebrospinal fluid, tend to mask identification and discovery of other less abundant proteins in two dimensional gel electrophoresis and other studies. AlbuminOUT™ has been specifically developed for substantial removal of albumin from such samples.
  • Rapid spin column format for albumin removal in under 10 minutes
  • Binding of albumin with Cibacron™ Blue dye
  • High capacity blue dye binding resin for instantaneous binding and removal of albumin
  • Column capacity: >2 mg human albumin per column
The albumin removal method is based on the binding of albumin with Cibacron Blue dye. AlbuminOUT™ has been optimised for removal of human albumin from samples. AlbuminOUT™ uses a rapid spin column method, where each column contains 0,2 ml dye-bound resins with capacity of greater than 2 mg human albumin per column. AlbuminOUT™ will remove over 98% albumin from 5 to 50 μl human plasma. The spin column format allows removal of albumin within ten minutes.
The high capacity blue dye-binding resin allows instantaneous binding and removal of albumin from human, pig, sheep, dog, rabbit, rat, and bovine samples. AlbuminOUT™ may also be used for removal of albumin from other species.
Order Now
Specificaties
Environmentally Preferable