Affinity purification resins, Glutathione Resin
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-310 786-311 786-312 786-280 786-541 786-540 786-587 786-588
786-310EA 1040 EUR
786-310 786-311 786-312 786-280 786-541 786-540 786-587 786-588
Affinity purification resins, Glutathione Resin
Eiwitzuivering Protein Purification Resins
Glutathione resin is designed for the affinity purification of proteins with a glutathione S-transferase (GST) tag. The resin consists of reduced glutathione (GSH) covalently coupled to 4% cross-linked agarose, via a 10-carbon spacer arm. The resin has a binding capacity of ~8 mg GST/ml resin. Supplied as slurry in 20% ethanol.
  • For the purification of GST proteins
  • High capacity (~8 mg/ml)
  • Ligand Density: 7 to 15 μmoles glutathione/ml
  • Bead size: 50 to 160 μm
  • Bead structure: 4% cross-linked agarose
  • 10 carbon spacer arm