Comfort microplate racks for upright freezers
Leverancier: Tenak
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TENATE35536EA 404.27 EUR
TENATE35536 TENATE35458 TENATE35414 TENATE35612 TENATE35416 TENATE35614 TENATE35422 TENATE35466 TENATE35586 TENATE35542 TENATE35424 TENATE35468 TENATE35500 TENATE35544 TENATE35462 TENATE35582 TENATE35420 TENATE35584 TENATE35540 TENATE35460 TENATE35580 TENATE35506 TENATE35508 TENATE35426 TENATE35502 TENATE35548 TENATE35598 TENATE35510 TENATE35432 TENATE35556 TENATE35434 TENATE35594 TENATE35550 TENATE35552 TENATE35596 TENATE35430 TENATE35590 TENATE35592 479-2130 TENATE35470 479-2132 479-2131 479-2134 479-2133 TENATE35558 TENATE35514 TENATE35488 TENATE35400 TENATE35520 TENATE35564 TENATE35402 TENATE35600 TENATE35522 TENATE35440 TENATE35484 TENATE35560 TENATE35442 TENATE35480 TENATE35482 479-2192 479-2191 TENATE35606 TENATE35404 TENATE35602 TENATE35604 TENATE35406 TENATE35526 TENATE35576 TENATE35532 TENATE35410 TENATE35456 TENATE35610 TENATE35412 TENATE35450 TENATE35530 TENATE35574 TENATE35452 479-2194 TENATE35490 479-2193 TENATE35570 479-2195
Comfort microplate racks for upright freezers
Rekken Rekken voor microplaten
This is a complete range of racks for microtiterplates for use in chest freezers.
Choose from 5 different shelf heights depending on preferences. The racks can either be used as one plate per shelf or the plates can be stacked. The racks are available with a shelf height of 16 mm, 25 mm, 45 mm, 65 mm and 85 mm. The external footprint is 135×92 mm on all racks. The comfort rack has an alert preventing the shelves from sliding out.