Filter holders, Swinnex®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SX0002500 SX0001300 SX0004700
511-0852EA 230 EUR
511-0852 511-0851 511-0853
Filter holders, Swinnex®
Filterhouders
These filter holders are used to ultraclean or sterilise small volumes of liquids dispensed by syringes.
47 mm filter holder connected with tubing to pressurised reservoir or automatic filling machine for filtering larger volumes.
Order Now
Specificaties
Filtratieoppervlak 3,4 cm² 0,7 cm² 13,8 cm²
Environmentally Preferable
In-/uitlaataansluiting verbindingen Female Luer lock®/male luer slip Female Luer lock®/male luer slip 6,35 mm NPTM /6,35 mm hose connector, with interior female luer slip