HPLC columns, Acclaim™ 120 C8
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
068970 069688 069696 071979 059122 059123 059124 059127 059134 059136 059138 059139 059140 059141
555-7140EA 949 EUR
555-7140 555-7115 555-7130 555-7179 555-7235 555-7236 555-7237 555-7240 555-7241 555-7243 555-7245 555-7246 555-7247 555-7248
HPLC columns, Acclaim™ 120 C8
Kolommen voor chromatografie
The Acclaim™ 120 C8 high performance reversed-phase columns are ideal for the separation of small molecules and this can be used for a variety of applications in the pharmaceutical, environmental, food and many other industrial sectors.
  • Low silanol activity for excellent peak shapes for basic analytes
  • Reproducible column-to-column performance
  • Extremely Low bleed, fully compatible with MS