Opticap® XL Filtration Capsules, Durapore® Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KVGLA10HH1 KVGLA04TT3 KVGLA02HH3 KVGLA10TT1 KVGLA02FF3 KVGLA04FH3 KVVLA05FF1 KVGLA02TT3
MILXKVGLA10HH1EA 843 EUR
MILXKVGLA10HH1 MILLKVGLA04NN3 MILXKVGLA04TT3 MILXKVGLA02HH3 MILXKVGLA10TT1 MILXKVGLA02FF3 MILXKVGLA04FH3 MILXKVVLA05FF1 MILXKVGLA02TT3
Opticap® XL Filtration Capsules, Durapore® Membrane
Filters Membraanfiltercapsules
Filtration capsules with PP housing, vent material and support material with double O ring seal.
  • Suitable for laboratory filtration
  • Aqueous extraction contains ≤0,5 EU/ml as determined by the Limulus Amebocyte Lysate (LAL) Test
  • The extractables level was equal to or less than 10 mg per capsule after 24 hours in water at controlled room temperature
  • It will meet the USP Oxidizable Substances Test requirements after a water flush of ≥500 ml
Opgelet: * Inlet temperature and pressure.