Afvoerschijven voor membraanfilters, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
230300 230600 231100 230500 230800
515-2340EA 71.1 EUR
515-2340 515-2342 515-2345 515-2341 515-2343
Afvoerschijven voor membraanfilters, Whatman™
Filters Filterschijven
The polyester drain disc is binder free and has a thickness of 100 µm.
  • Flat surface to eliminate filter tearing or rupturing
  • Used as a separator between membrane layers in serial stack filtration applications
This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.