Gel filtration calibration kits
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-4038-42 28-4038-41
28-4038-42EA 677 EUR
28-4038-42 28-4038-41
Gel filtration calibration kits
Eiwitzuivering Protein Purification Resins
Gel Filtration Calibration Kits are suitable for calibration of high-resolution gel filtration media and columns, such as Superdex, Superose and Sephacryl.
  • Optimised range of proteins suit high resolution media and pre-packed columns
  • Low and high molecular weight ranges from 6500 to 75000 and 43000 to 669000 respectively
  • Each kit includes five proteins lyophilised in individual vials
  • Includes Blue Dextran 2000 to determine the void fraction in the column (also available separately)
Opgelet: For research use only.