Mini desiccator, SICCO Mini Secure Box
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V1847-06 V1947-06
BOHLV1847-06EA 293 EUR
BOHLV1847-06 BOHLV1947-06
Mini desiccator, SICCO Mini Secure Box
Exsiccatoren Exsiccatorkasten
Clear polycarbonate lockable desiccators with single door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight, portable and stackable
  • Premium model includes hygrometer and two shelves
Leveringsinformatie: Wordt geleverd met droogmiddel in een handige hersluitbare container.