Mini desiccator for gas filling, SICCO Mini
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V1849-06 V1949-06
BOHLV1849-06EA 459 EUR
BOHLV1849-06 BOHLV1949-06
Mini desiccator for gas filling, SICCO Mini
Exsiccatoren Exsiccatorkasten
Clear polycarbonate desiccators for gas filling with single door with snap fit closing and integrated gasket ensuring easy opening and tight closure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight and stackable
  • Premium model includes hygrometer and two shelves
Unit has tubing with quick couplings for gas filling. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
Leveringsinformatie: Supplied with desiccant which is in a handy, resealable container.