Watch dishes
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113197 113297 113397 113497
VITL113197EA 14.8 EUR
VITL113197 VITL113297 VITL113397 VITL113497
Watch dishes
Horlogeglazen
Watch dishes, made of PTFE, white.
  • Without base
  • Very good chemical resistance
  • Good temperature resistance
Ideal for use to cover beakers.
Certificaten: DIN EN 285
Order Now
Specificaties
Environmentally Preferable