Affinity chromatography media, TALON® Superflow™
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-9574-99 28-9575-02
28-9574-99EA 261 EUR
28-9574-99 28-9575-02
Affinity chromatography media, TALON® Superflow™
Eiwitzuivering Protein Purification Resins
Cobalt-based chromatography medium designed for purification of histidine-tagged recombinant proteins by immobilised metal affinity chromatography (IMAC).
  • Suitable when Ni²⁺ is not the optimal choice of metal ion
  • Compatible with commonly used IMAC reagents
  • Allows protein purification under native or denaturing conditions
  • Used with prokaryotic and eukaryotic expression systems