Bottle top vacuum filtration systems
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5809/2 5809/3 5810/5 5810/3 5810/4 5809/KIT
514-0698EA 120 EUR
514-0698 BIBB5809/3 514-0701 514-0699 514-0700 514-0697
Bottle top vacuum filtration systems
Filters Flesfilters
This filteration system is used for microbiology and preparation of HPLC solvents. It is used with 47 mm Ø membranes.
  • Borosilicate glass sintered disc to support membrane
  • Each component is also available to order separately
Order Now
Specificaties
Environmentally Preferable