InGel™ for protein digestion
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-241EA 528 EUR
786-241
InGel™ for protein digestion
Eiwitextractie Reagentia voor eiwitextractie
The digested peptides are extracted with Pep-Extract™, a high diffusion peptide extraction buffer. The extracted peptides are suitable for mass spectrometry analysis without any subsequent treatments or cleaning procedures.
Leveringsinformatie: The InGel™ kit is supplied with all the necessary reagents for 100 protein spots and includes: SilverOUT™ destaining reagent; MSG-Trypsin™, a mass spectrometry grade trypsin with minimal autolysis and increases stability; trypsin digestion buffer, for optimal trypsin activity; and Pep-Extract™ Buffer, for high level peptide extraction.