PIG® barrel top mat
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT208 MAT244
NEWPMAT208EA 107 EUR
NEWPMAT208 NEWPMAT244
PIG® barrel top mat
Absorptiematerialen en bescherming tegen morsen
Circular-shaped PIG mat is pre-cut for a perfect fit on drum tops. Keep messy pump drips in check for clean, safe dispensing areas.
  • Soak up pump drips without cluttering up your work space with dirty rags
  • Eight layers of 100% polypropylene are thermally bonded; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime, so mat stays on the job longer
  • Fits 210 L steel drum tops; pre-cut with holes