Fluorescent labelling reagents, CypHer5E NHS Ester
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PA15401 PA15405
PA15401EA 531 EUR
PA15401 PA15405
Fluorescent labelling reagents, CypHer5E NHS Ester
Reagentia voor elektroforese Western Blotting Reagents
CypHer5E is a red-excitable, pH-sensitive cyanine dye derivative that is minimally fluorescent at a basic pH and maximally fluorescent at an acidic pH.
  • Generic technology can be coupled to any receptor or epitope tag antibody
  • Monitors any neutral to acidic event
  • Simple fluorescence intensity increase
It is well suited to report the movement of a receptor from the cell surface into acidic endosomes upon agonist stimulation.