Chemiluminescent detection reagents, Immobilon® Western HRP substrates
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WBKLS0100 WBKLS0500 WBKLS0050
MILFWBKLS0100EA 171 EUR
MILFWBKLS0100 MILFWBKLS0500 MILFWBKLS0050
Chemiluminescent detection reagents, Immobilon® Western HRP substrates
Reagentia voor elektroforese Western Blotting Reagents
Immobilon® Western HRP substrates are compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.