Centrifugal filter units, Amicon® Ultra-15
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UFC901008 UFC901024 UFC901096 UFC903008 UFC903024 UFC903096 UFC910008 UFC910024 UFC910096 UFC905008 UFC905024 UFC900308 UFC900324 UFC905096 UFC900396
516-0555EA 145 EUR
516-0555 516-0556 MILFUFC901096 516-0557 MILFUFC903024 MILFUFC903096 516-0559 516-0560 MILFUFC910096 516-0627 516-0628 MILFUFC900308 MILFUFC900324 MILFUFC905096 MILFUFC900396
Centrifugal filter units, Amicon® Ultra-15
Filters Centrifugefilters
These centrifugal filter units can be used for a number of applications: concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; purification of macromolecular components found in tissue culture extracts or cell lysates and protein removal prior to HPLS; concentration of dilute or pre-purified proteins from column eluents; desalting, buffer exchange and protein dialysis; size exclusion and protein fractionation.
  • High retentate recovery of >90%; 100% integrity tested
  • Vertical membrane reduces concentration polarisation for ultra-fast spin times
  • Heat-sealed membrane minimises downstream extractables
  • Convenient sample monitoring with translucent housing and volume graduations
  • Direct pipettor sample access eliminates processing step to recover concentrate
  • High concentration factors of 80 - 100X
Membrane: Ultracel regenerated cellulose; housing: styrene/butadiene; tube: PP; cap and liner: PE.