Ficoll® PM70
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17-0310-05 17-0310-10 17-0310-50
17-0310-05EA 6515 EUR
17-0310-05 17-0310-10 17-0310-50
Ficoll® PM70
Media voor celscheiding
Ficoll® PM70 is a high molecular weight sucrose polymer formed by copolymerisation of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
  • Displays minimal toxicity
  • Favourable viscosity and osmotic properties
The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.