Spray bottles
Leverancier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P01422-P51314 P014/22P513/14
MURRP01422-P51314EA 461 EUR
MURRP01422-P51314 MURRP014/22P513/14
Spray bottles
Flessen Flessen met verstuiver
HDPE, translucent.
Designed to allow comfortable grip and minimise fatigue. Adjustable from fine spray to single jet. Delivery 1,4 ml per stroke.