Bowls, PP
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42594 42694 42894 42794 42994 43094 43194
216-1201EA 2.9 EUR
216-1201 216-1202 216-1204 216-1203 216-1205 216-1206 216-1207
Bowls, PP
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Bowls, made of white PP.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
Ideal for safe storage of food products. With broad, stable edge and circumferential standing ring on the bottom.