Blood collection tubes, without additive, VACUETTE®
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
454001_1200 455001_1200 456001_1200
GREI454001_1200EA 196 EUR
GREI454001_1200 GREI455001_1200 GREI456001_1200
Blood collection tubes, without additive, VACUETTE®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, clear.
  • With writing area
  • With ridged safety twist cap or non-ridged pull cap
The VACUETTE® blood collection tubes made from practically unbreakable plastic have become well established on the market. They have made the drawing of blood a safer and more hygienic process. The use of PET plastic offers the advantage that the tubes are as clear as glass.