Oil sorbents mats
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11327 9934
MSGA11327EA 144 EUR
MSGA11327 MSGA9934
Oil sorbents mats
Absorptiematerialen en bescherming tegen morsen
Water repellent mats for abosrption and clear up of oils and petrol based liquid products.
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Perforated to only take sufficient material as required