Oil sorbents socks
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9930 9931
MSGA9930EA 90 EUR
MSGA9930 MSGA9931
Oil sorbents socks
Absorptiematerialen en bescherming tegen morsen
Water repellent absorbent socks for safe absorption of oil and petrol.
  • Creates an absorption barrier to enclose larger leaks
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance