Universal absorbent pillows
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9943 9944
MSGA9943EA 121 EUR
MSGA9943 MSGA9944
Universal absorbent pillows
Absorptiematerialen en bescherming tegen morsen
Universal pillows for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for those hard to reach places
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance