Absorbent pillows
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9910 9916
MSGA9910EA 148 EUR
MSGA9910 MSGA9916
Absorbent pillows
Absorptiematerialen en bescherming tegen morsen
Special pillows for safe absorption of acids, alkalis, aggressive chemicals and oil.