CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Leverancier: BIOCISION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BCS-206B BCS-206 BCS-206G BCS-206O BCS-206P BCS-206PK BCS-207 BCS-207B BCS-207G BCS-207O BCS-207P BCS-207PK BCS-206MC BCS-217G BCS-217P BCS-221G BCS-221P BCS-209G BCS-209P BCS-220G BCS-220P BCS-215G BCS-215P BCS-219G BCS-219P
479-1351EA 117 EUR
479-1351 479-1350 479-1352 479-1354 479-1355 479-1356 479-1357 479-1358 479-1359 479-1360 479-1361 479-1362 479-1353 479-1892 479-1893 479-1894 479-1895 479-1363 479-1364 479-1365 479-1366 479-1371 479-1372 479-1373 479-1374
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Dozen Dozen voor cryobuisjes
Laminated cardboard.
  • Designed for repeated freezing and thawing in vapor-phase liquid nitrogen
  • Standard boxes available in 6 colours
  • Available in 2 heights, 5,2 cm and 9,2 cm to accommodate 1 to 5 ml cryotubes
TruCool® Cryoboxes have a hinged lid that remains attached which ensures lid is placed on correctly and markings are in sync with samples. Lid is easy to open, even when frozen and stays attached to the base minimising risk of contamination. Boxes have a white patch on the front for markings or barcode. Made from laminated cardboard, waterproof and durable. Includes 81-place adjustable plastic grid or 100-place plastic grid which can be removed if needed. Dual indexing system: 1 to 81 grid on the top and inside cover, A to I and 1 to 9 column and row indexing around the inner and outer edge of box.
Dimensions: 133,3×133,3×51 mm or 133,3×133,3×92 mm (L×W×H).
Dimensions: 133,3×133,3×51 mm or 133,3×133,3×92 mm (L×W×H)